Learn More
LPAM-1 Rat anti-Mouse, Brilliant Violet 786, Clone: DATK32, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740892
Description
The DATK32 monoclonal antibody specifically binds to an epitope specific for the α 4β 7 integrin heterodimer. α 4β 7 (LPAM-1) is expressed on most mature lymphocytes and on small subsets of thymic and bone marrow cells. It interacts with several ligands, including VCAM-1 (CD106), fibronectin, and MAdCAM-1. DATK32 antibody induces α 4β 7-dependent lymphocyte aggregation, but it inhibits other α
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).

Specifications
LPAM-1 | |
Monoclonal | |
0.2mg/mL | |
Itga4Itgb7; α4β7 Integrin; alpha 4 beta 7 integrin; CD49dβ7 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
DATK32 | |
Brilliant Violet 786 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
AKR/Cum mouse T lymphoma line TK1 | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only