Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6G and Ly-6C Rat anti-Mouse, Brilliant Violet 650, Clone: RB6-8C5, BD Optibuild™

Catalog No. BDB740454 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740454 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740454 Supplier BD Biosciences Supplier No. 740454
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The RB6-8C5 monoclonal antibody recognizes a common epitope on Ly-6G and Ly-6C, previously known as the myeloid differentiation antigen Gr-1. In the bone marrow, the level of antigen expression is directly correlated with granulocyte differentiation and maturation. The antigen is also expressed on the monocyte lineage in the bone marrow, but not on erythroid cells. In the periphery, RB6-8C5 antibody recognizes granulocytes (neutrophils and eosinophils) and monocytes. The RB6-8C5 antibody is a component of the “lineage cocktail” used in studies of hematopoietic cell lineages. The 1A8 antibody (Cat. No. 551461) specifically recognizes Ly-6G, but not Ly-6C.

Based on comparison of the staining patterns given by 1A8 versus RB6-8C5 antibodies on total blood leucocytes, it is evident that the 1A8 antibody stains the RB6-8C5-bright population, corresponding to Ly-6G-expressing granulocytes; whereas, the RB6-8C5-dim population is 1A8-negative and corresponds to Ly-6C-expressing lymphocytes and monocytes. Please refer to the Technical Data Sheets for Cat. No. 551459 and 553128 for more detailed information.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6G and Ly-6C
Applications Flow Cytometry
Classification Monoclonal
Clone RB6-8C5
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Description Ly6c, Lymphocyte antigen 6C2; Lymphocyte antigen 6G, Ly6g, Gr-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Not Reported
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.