Learn More
Ly-6G and Ly-6C Rat anti-Mouse, Brilliant Violet 786, Clone: RB6-8C5, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740850
Description
The RB6-8C5 monoclonal antibody recognizes a common epitope on Ly-6G and Ly-6C, previously known as the myeloid differentiation antigen Gr-1. In the bone marrow, the level of antigen expression is directly correlated with granulocyte differentiation and maturation. The antigen is also expressed on the monocyte lineage in the bone marrow, but not on erythroid cells. In the periphery, RB6-8C5 antibody recognizes granulocytes (neutrophils and eosinophils) and monocytes. The RB6-8C5 antibody is a component of the “lineage cocktail” used in studies of hematopoietic cell lineages. The 1A8 antibody (Cat. No. 551461) specifically recognizes Ly-6G, but not Ly-6C.
Based on comparison of the staining patterns given by 1A8 versus RB6-8C5 antibodies on total blood leucocytes, it is evident that the 1A8 antibody stains the RB6-8C5-bright population, corresponding to Ly-6G-expressing granulocytes; whereas, the RB6-8C5-dim population is 1A8-negative and corresponds to Ly-6C-expressing lymphocytes and monocytes. Please refer to the Technical Data Sheets for Cat. No. 551459 and 553128 for more detailed information.

Specifications
Ly-6G and Ly-6C | |
Monoclonal | |
0.2mg/mL | |
Ly6c, Lymphocyte antigen 6C2; Lymphocyte antigen 6G, Ly6g, Gr-1 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
RB6-8C5 | |
Brilliant Violet 786 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Not Reported | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only