Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MVP, Rat, Clone: LMR5, Abnova™

Catalog No. 89135508 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-508 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-508 Supplier Abnova Corporation Supplier No. MAB3733
Add to Cart
Add to Cart

Rat monoclonal antibody raised against MVP.

Specifications

Antigen MVP
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone LMR5
Conjugate Unconjugated
Description Rat monoclonal antibody raised against MVP.
Dilution Flow Cytometry (1:50) Immunohistochemistry (Frozen sections) (1:20-1:50) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:20) The optimal working dilution should be determined by the end user.
Formulation In serum-free culture supernatant (0.09% sodium azide)
Host Species Rat
Immunogen MVP (110 kDa).
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Monoclonal antibody recognizes the 110 KDa Lung Resistance Protein (LRP). Reacts with an internal epitope of the LRP-protein (p110). LRP, also known as the Major Vault Protein, is upregulated and strongly overexpressed in various human P-glycoprotein negative MDR tumor cells. Can be used for the detection of LRP-associated non-Pglycoprotein MDR in human tumor samples.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.