Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nes, Rat, Clone: 7A3, Abnova™

Catalog No. 89113031 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-031 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-031 Supplier Abnova Corporation Supplier No. MAB6767

Rat monoclonal antibody raised against native Nes.

Specifications

Antigen Nes
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 7A3
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native Nes.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol)
Gene Nes
Gene Alias AA166324/C78523/ESTM46
Gene Symbols Nes
Host Species Rat
Immunogen Native Nes from mouse E16 embryonic cerebral cortex extracts.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18008
Test Specificity This antibody is specific to mouse Nes.
Target Species Mouse
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.