Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nid1, Rat, Clone: JF2, Abnova™

Catalog No. 89111008 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-008 Supplier Abnova Corporation Supplier No. MAB2454
Add to Cart
Add to Cart

Rat monoclonal antibody raised against partial recombinant Nid1.

Specifications

Antigen Nid1
Applications ELISA, Western Blot
Classification Monoclonal
Clone JF2
Conjugate Unconjugated
Description Rat monoclonal antibody raised against partial recombinant Nid1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In culture supernatant
Gene Nid1
Gene Alias A630025O17/Nid/entactin/entactin-1/nidogen-1
Gene Symbols Nid1
Host Species Rat
Immunogen Recombinant protein corresponding to G3 region of mouse Nid1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18073
Test Specificity recognizes native and denatured mouse and human Nid1, and blocks binding of Nid1 to laminin.
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.