Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nid1, Rat, Clone: JF4, Abnova™
Rat monoclonal antibody raised against partial recombinant Nid1.
Supplier: Abnova Corporation MAB2456
Specifications
Nid1 | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Nid1 | |
Nid1 | |
Recombinant protein corresponding to G2 region of mouse Nid1. | |
RUO | |
18073 | |
Human, Mouse | |
Liquid |
ELISA, Western Blot | |
JF4 | |
Rat monoclonal antibody raised against partial recombinant Nid1. | |
In culture supernatant | |
A630025O17/Nid/entactin/entactin-1/nidogen-1 | |
Rat | |
100 μL | |
Primary | |
recognizes native and denatured mouse and human Nid1, and blocks binding of Nid1 to laminin. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction