Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NO-L-Cysteine, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89136846 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-846 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-846 Supplier Abnova Corporation Supplier No. PAB0064

Rat polyclonal antibody raised against NO-L-Cysteine.

Specifications

Antigen NO-L-Cysteine
Applications ELISA, Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Rat polyclonal antibody raised against NO-L-Cysteine.
Dilution ELISA (1:2000-1:10000) Immunohistochemistry (1:1000-1:5000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.4
Host Species Rat
Immunogen NO-L-Cysteine conjugated with carrier protein.
Purification Method Ammonium sulfate precipitation
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 50μL of distilled water and 50μL of glycerol, store at -20°C for 2 years. Aliquot to avoid repeated freezing and thawing.
Product Type NO-L-Cysteine polyclonal antibody
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.