Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nup153, Rat, Clone: R4C8, Abnova™

Catalog No. 89113024 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-024 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-024 Supplier Abnova Corporation Supplier No. MAB6759
Add to Cart
Add to Cart

Rat monoclonal antibody raised against partial recombinant Nup153.

Specifications

Antigen Nup153
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone R4C8
Conjugate Unconjugated
Description Rat monoclonal antibody raised against partial recombinant Nup153.
Dilution The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.05% sodium azide)
Gene Nup153
Gene Alias NUCZINK
Gene Symbols Nup153
Host Species Rat
Immunogen Recombinant GST fusion protein corresponding to amino acids 610-1191 rat Nup153.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25281
Target Species Human, Monkey, Mouse, Rat
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.