Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rat anti-Nup96, Clone: 4H5,

Catalog No. MABE1039MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1039MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1039MI Supplier MilliporeSigma™ Supplier No. MABE1039
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects Nup96 clone: 4H5 in Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

The murine gene Nup98 (Gene ID 269966; also known as AI849286, 4732457F17) encodes the 1816-amino acid nuclear pore complex protein Nup98-Nup96 (UniProt Q6PFD9), which is posttranslationally autocleaved between aa 800 and 801 to form Nup98 (aa 1-800) and Nup96 (aa 801-1816). The nuclear pore complex (NPC) acts as a passageway for nucleocytoplasmic transport. The core structure of the NPC is maintained by the Nup93-205 complex sandwiched between the supporting structures of the Nup107-160 complex on the nuclear and cytoplasmic faces of the pore. Nup96 is a component of the Nup107-160 complex, the largest subunit of the NPC. Nup96 knockout is reported to be fatal in mice and Nup96(±) mice with low levels of Nup96 display impaired immune response and are highly susceptible to viral infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nup96
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 4H5
Formulation Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q6PFD9
Gene Symbols Nup98
Host Species Rat
Immunogen GST-tagged recombinant protein corresponding to mouse Nup96 near the N-terminus.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001274093
Target Species Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.