Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rat anti-Nup98, Clone: 2H10,

Catalog No. MABE1040MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1040MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1040MI Supplier MilliporeSigma™ Supplier No. MABE1040
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects Nup98 clone: 2H10 in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Western Blotting

The human gene NUP98 (Gene ID 4928; Also known as ADAR2, ADIR2, NUP96, NUP196) encodes the nuclear pore complex protein Nup98-Nup96 (UniProt P52948; also known as GLFG-repeat containing nucleoporin, Nup98-Nup96), which is posttranslationally autocleaved to form Nup98 and Nup96. Cleavage of the Nup98-96 linkage enhances the interaction between Nup98 and Nup96 and promotes localization of the two proteins to the nuclear pore complex (NPC). An alternative splice event generates a mRNA variant (isoform 3; also known as Nup98-specific 1), which expresses only the Nup98 portion of the Nup98-96 mRNA and allows the independent production of Nup98. Unlike Nup96, Nup98 is known to exist in both NPC-bound and non-NPC-associated pool, where it may participate in gene transcription regulations. Nup98 is reported to interact with histone-modifying enzymes CBP/p300 and histone deacetylases (HDACs) through its glycine-leucine-phenylalanine-glycine (GLFG) repeats. NUP98 gene is also involved in chromosomal translocations, resulting in Nup98 translocation fusions identified as potent transcription factors that induce acute myeloid leukemia (AML) with a particularly poor patient prognosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nup98
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2H10
Formulation Purified rat monoclonal IgG2cκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P52948
Gene Symbols NUP98; ADAR2
Host Species Rat
Immunogen GST-tagged recombinant protein corresponding to human Nup98 near the N-terminus.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_005378
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2c κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.