Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oval cell marker, Rat, Clone: OC2-3C5, Abnova™

Catalog No. 89136346 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-346 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-346 Supplier Abnova Corporation Supplier No. MAB7742
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native oval cell marker.

Specifications

Antigen Oval cell marker
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OC2-3C5
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native oval cell marker.
Dilution Flow Cytometry (1:50-1:100) Immunohistochemistry (Frozen sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant (0.05% sodium azide)
Host Species Rat
Immunogen Native purified oval cell marker from nonparenchymal cells from DDC-treated mice to induce the oval cell response.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for both proliferating and normal hepatic periductal and ductal cells.
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.