Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rat anti-Pax-9, Clone: 7C2 (P9C23),

Catalog No. MABE1041MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1041MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1041MI Supplier MilliporeSigma™ Supplier No. MABE1041
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects Pax-9 clone: 7C2 (P9C23) in Human, Mouse samples, and it is validated for Western Blotting

Paired box protein Pax-9 (UniProt P47242; also known as Paired box gene 9) is encoded by the Pax9 gene (Gene ID 18511) in murine species. Pax9 belongs to a family of nine transcription factors (Pax1-9) characterized by the presence of a DNA-binding ‘paired’-domain. Pax family transcription factors regulate the morphogenesis of a wide range of organs and are key factors for the development of mammalian sensory organs such as the eye (Pax6, Pax2), nose (Pax6), ears (Pax2, Pax8), and the endoderm-derived taste papillae (Pax9). In addition, Pax9 expression in the normal epithelium of the adult human oesophagus is reported to be either lost or significantly reduced in the majority of invasive carcinomas and epithelial dysplasias (precancerous lesions). The drop in the percentage of Pax9-positive cells in the epithelium decreases with increasing malignancy of the epithelial lesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pax-9
Applications Western Blot
Classification Monoclonal
Clone 7C2 (P9C23)
Formulation Purified rat monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P47242
Gene Symbols Pax9; Pax-9
Host Species Rat
Immunogen MBP-tagged recombinant protein corresponding to the C-terminal region of mouse Pax-9.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_035171
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.