Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGFR alpha Antibody (APA5) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$340.50 - $499.50
Specifications
Antigen | PDGF R alpha |
---|---|
Clone | APA5 |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14335005
![]() |
Novus Biologicals
NBP1433500.05MG |
0.05 mg |
Each for $340.50
|
|
|||||
NBP14335001
![]() |
Novus Biologicals
NBP1433500.1MG |
0.1 mg |
Each for $499.50
|
|
|||||
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
PDGF R alpha | |
0.5 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Unconjugated | |
RUO | |
Mouse | |
P26618 | |
5156 | |
The immunogen for this antibody was CD140a. | |
Primary | |
Store at 4C. Do not freeze. |
APA5 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Rat | |
Protein Kinase | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
PDGFRA | |
IgG2a κ | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title