Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pdgfrb, Rat, Clone: APB5, Abnova™

Catalog No. 89109181 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-181 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-181 Supplier Abnova Corporation Supplier No. MAB10966

Rat monoclonal antibody raised against Pdgfrb.

Specifications

Antigen Pdgfrb
Applications Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone APB5
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Pdgfrb.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Gene Pdgfrb
Gene Alias AI528809/CD140b/Pdgfr
Gene Symbols Pdgfrb
Host Species Rat
Immunogen Mouse Pdgfrb.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18596
Test Specificity The monoclonal antibody reacts with the mouse Pdgfra, the beta chain of the platelet derived growth factor receptor (PDGF receptor).
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.