Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pecam1, Rat, Clone: MEC13.3, Abnova™

Catalog No. 89113127 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-127 Supplier Abnova Corporation Supplier No. MAB7099
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Pecam1.

Specifications

Antigen Pecam1
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone MEC13.3
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Pecam1.
Dilution Flow Cytometry (1:1-1:10) Immunocytochemistry (1:1-1:10) Immunohistochemistry (Frozen sections) (1:1-1:10) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1-1:10) Immunoprecipitation (1:1-1:10) The optimal working di
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Gene Pecam1
Gene Alias C85791/Cd31/MGC102160/PECAM-1/Pecam
Gene Symbols Pecam1
Host Species Rat
Immunogen Full length mouse Pecam1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18613
Test Specificity CD31 (MEC13.3).
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.