Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pecam1, Rat, FITC, Clone: 390, Abnova™

Catalog No. 89112385 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-385 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-385 Supplier Abnova Corporation Supplier No. MAB5712
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native Pecam1.

Specifications

Antigen Pecam1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 390
Conjugate FITC
Description Rat monoclonal antibody raised against native Pecam1.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Pecam1
Gene Alias C85791/Cd31/MGC102160/PECAM-1/Pecam
Gene Symbols Pecam1
Host Species Rat
Immunogen Native purified Pecam1 from mouse leukocyte cell line 32D
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18613
Test Specificity Mouse CD31/PECAM-1, Mr 130-140 KDa.
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.