Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHLDA3, Rat, Clone: 4B6, Abnova™

Catalog No. 89113021 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-021 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-021 Supplier Abnova Corporation Supplier No. MAB6755
Add to Cart
Add to Cart

Rat monoclonal antibody raised against synthetic peptide of PHLDA3.

Specifications

Antigen PHLDA3
Applications Western Blot
Classification Monoclonal
Clone 4B6
Conjugate Unconjugated
Description Rat monoclonal antibody raised against synthetic peptide of PHLDA3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol)
Gene PHLDA3
Gene Alias TIH1
Gene Symbols PHLDA3
Host Species Rat
Immunogen A synthetic peptide corresponding to amino acids 1-31 of human PHLDA3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23612
Test Specificity This antibody is specific to human PHLDA3.
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.