Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proc Rat anti-Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89107907 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-907 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-907 Supplier Abnova Corporation Supplier No. MAB0282
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native Proc.

Specifications

Antigen Proc
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native Proc.
Dilution Western Blot (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol)
Gene Proc
Gene Alias PC
Gene Symbols Proc
Host Species Rat
Immunogen Native purified Mouse Proc.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19123
Test Specificity This antibody recognizes mouse protein C.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.