Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rat anti-Progranulin/GRN, Clone: 8H10,

Catalog No. MABN1795MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1795MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1795MI Supplier MilliporeSigma™ Supplier No. MABN1795
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects Progranulin/GRN clone: 8H10 in Mouse samples, and it is validated for Immunohistochemistry, Western Blotting

Granulins constitute a family of secreted, glycosylated peptides that are cleaved from a single precursor protein with 7.5 repeats of a highly conserved 12-cysteine granulin/epithelin motif. The 602 aa murine precursor protein, preprogranulin (UniProt Q3U9N4), is also called proepithelin and PC cell-derived growth factor. Cleavage of the signal peptide (aa 1-17) produces progranulin (Acrogranin; aa 18-589), which can be further cleaved into 7 active peptides named granulin-1 through granulin-7. Both the peptides and progranulin protein regulate cell growth. Granulin family members are important in normal development, wound healing, and tumorigenesis. Heterozygous loss-of-function mutations in the GRN gene is a known cause for frontotemporal lobar degeneration (FTLD) with accumulation of TAR DNA-binding protein (TDP)-43, while a complete GRN deficiency due to a homozygous loss-of-function mutation causes the lysosomal storage disorder known as neuronal ceroid lipofuscinosis (NCL). In Grn-/- mice, a murine model for GRN-associated FTLD-TDP (FTLD-TDP/GRN), the lysosomal proteins cathepsin D (CTSD) is found to be elevated, while an upregulated granulin level is observed in Ctsd(-/-) mice, a model for NCL. Moreover, both types of knockout mice are found to display increased levels of transmembrane protein 106B (TMEM106B) as a common phenotype.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Progranulin/GRN
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8H10
Formulation Purified rat monoclonal IgG2cκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q3U9N4
Gene Symbols Grn
Host Species Rat
Immunogen Linear peptide corresponding to rat progranulin sequence near the C-terminus
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_032201
Target Species Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2c κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.