Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rat anti-Prohibitin-2/PHB2, Clone: 7F8E3,

Catalog No. MABC953MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC953MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC953MI Supplier MilliporeSigma™ Supplier No. MABC953
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects Prohibitin-2/PHB2 clone: 7F8E3 in Human, Monkey, Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting

Prohibitin-2 (UniProt Q99623; also known as B-cell associated protein, B-cell receptor-associated protein BAP37, BCAP37, D-prohibitin, PHB2, Repressor of estrogen receptor activity) is encoded by the PHB2 (also known as BAP, REA) gene (Gene ID 11331) in human. Prohibitins are assembled into a ring-like PHB complex with 16-20 alternating Phb1 and Phb2 subunits in the inner mitochondrial membrane, where they function as chaperone for respiration chain proteins or as important structural scaffold for optimal mitochondrial morphology and function. Prohibitins are also reported to modulate transcriptions by interacting with various transcription factors, including nuclear receptors. Upregulated expression of prohibitins, septin 9, neurabin 1, and other cytoskeletal and functional proteins is found at the glutathione S-transferase placental-form-positive (GST-P+) hepatocellular carcinoma (HCC) foci during initiation and/or promotion stages of rat hepatocarcinogenesis, implicating an involvement of prohibitins in the neoplastic transformation of rat liver preneoplastic lesions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prohibitin-2/PHB2
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7F8E3
Formulation Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q99623
Gene Symbols PHB2; BAP; REA
Host Species Rat
Immunogen KLH-conjugated linear peptide corresponding to human Prohibitin-2/PHB2.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001138303
Target Species Human, Monkey, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.