Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pvrl3, Rat, Clone: 103-A1, Abnova™

Catalog No. 89112158 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-158 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-158 Supplier Abnova Corporation Supplier No. MAB5184
Add to Cart
Add to Cart

Rat monoclonal antibody raised against partial recombinant Pvrl3.

Specifications

Antigen Pvrl3
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 103-A1
Conjugate Unconjugated
Description Rat monoclonal antibody raised against partial recombinant Pvrl3.
Dilution Recommend dilution at 1:10 The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% BSA, 0.02% sodium azide)
Gene Pvrl3
Gene Alias 2610301B19Rik/3000002N23Rik/4921513D19Rik/AA407785/AU016832/AW538082
Gene Symbols Pvrl3
Host Species Rat
Immunogen Recombinant protein corresponding to amino acids 50-400 of mouse Pvrl3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58998
Test Specificity Nectin-3 also known as PRR3 is a transmembrane protein that is mainly expressed in testis and placental tissues. Nectin-3/PRR3 interacts in vivo with both long and short isoforms of afadin. Antibody 103-A1 does not react with human and rat nectin-3. Antibody 103-A1 cannot be used for Western blot.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.