Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sell, Rat, APC-Cy7, Clone: MEL-14, Abnova™

Catalog No. 89112413 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-413 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-413 Supplier Abnova Corporation Supplier No. MAB5741
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Sell.

Specifications

Antigen Sell
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone MEL-14
Conjugate APC-Cyanine7
Description Rat monoclonal antibody raised against Sell.
Dilution Flow Cytometry (0.1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Sell
Gene Alias AI528707/CD62L/L-selectin/LECAM-1/Lnhr/Ly-22/Ly-m22/Lyam-1/Lyam1
Gene Symbols Sell
Host Species Rat
Immunogen C3H/eb cloned mouse B lymphoma 38C-13.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20343
Test Specificity CD62L/L-selectin, Mr 90 KDa.
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.