Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rat anti-Semaphorin 4D/CD100, Clone: BMA-12,
Rat Monoclonal Antibody
Supplier: MilliporeSigma™ MABC987
Description
Specifically detects Semaphorin 4D/CD100 clone: BMA-12 in Mouse samples, and it is validated for Activity Assay, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blotting
Semaphorin 4D (alson known as CD100, Sema J, M-sema G, Semaphorin H, Semaphorin-J, Semaphorin-C-like 2) is a single-transmembrane protein encoded by the Sema4d gene in mouse. Semaphorins are involved in a wide range of biological processes, including axon guidance, neuronal migration, angiogenesis, cardio- and osteo-genesis, via interaction with a family of transmembrane proteins called Plexins expressed on other cells. Semaphroins and Plexins interact via their conserved sema domains, each sema domain forms a seven-blade beta-propeller fold in the extracellular domain of the respective protein. Semaphorin 4D extracellular domain can also be cleaved off and function in soluble form. Estrogen-stimulated Sema4D shedding is important for apoptosis induction of Plexin-B1-expressing vaginal epithelium during postnatal tissue remodeling (PMID 24841081). In addition, MT1-MMP-mediated Sema4D shedding is reported to induce endothelial cell chemotaxis in vitro and blood vessel growth in vivo and forms the basis of tumor-induced angiogenesis among several head and neck squamous cell carcinoma cell lines (PMID 17204469). Interference of cell communication between Sema4D-expressing osteoclasts and PlexinB1-expressing osteoblasts is reported to be therapeutically beneficial in a murine model of osteoporosis (PMID 25088728 & 25252878).
Specifications
Semaphorin 4D/CD100 | |
Monoclonal | |
Purified rat monoclonal IgG2a in buffer containing PBS without preservatives. | |
Sema4d; Semacl2; Semaj | |
Recombinant protein corresponding to mouse Semaphorin 4D/CD100. | |
100 μg | |
Apoptosis, Cancer | |
NP_001268809 | |
Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance. | |
IgG2a |
Functional Assay, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
BMA-12 | |
O09126 | |
Rat | |
Protein G Purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction