Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smc1a, Rat, Clone: KT73, Abnova™

Catalog No. 89108432 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-432 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-432 Supplier Abnova Corporation Supplier No. MAB10227
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant Smc1a.

Specifications

Antigen Smc1a
Applications Western Blot
Classification Monoclonal
Clone KT73
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant Smc1a.
Dilution Western Blot (1:100) The optimal working dilution should be determined by the end user.
Formulation In culture supernatant (0.09% sodium azide)
Gene Smc1a
Gene Alias 5830426I24Rik/KIAA0178/SB1.8/SMCB/Smc1/Smc1alpha/Smc1l1/mKIAA0178
Gene Symbols Smc1a
Host Species Rat
Immunogen Recombinant GST fusion protein corresponding to mouse Smc1a.
Quantity 3 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24061
Target Species Mouse
Content And Storage Store at 4°C for one month. For long term storage store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.