Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Substance P, Rat, Clone: NC1/34, Abnova™

Catalog No. 89136040 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-040 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-040 Supplier Abnova Corporation Supplier No. MAB5682
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Substance P.

Specifications

Antigen Substance P
Applications Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone NC1/34
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Substance P.
Dilution Immunohistochemistry (1:200) Immunocytochemistry (1:200) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Host Species Rat
Immunogen Substance P.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes COOH terminal end of substance P. It doesn't recognize Leu or Met enkephalin, somatostatin or beta endorphin; cross reactivity with eledoisin: 5%.
Target Species Mammalia
Content And Storage Store at -20°C. After reconstitution with 500μL distilled water, store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.