Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tek, Rat, Clone: TEK4, Abnova™

Catalog No. 89109187 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-187 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-187 Supplier Abnova Corporation Supplier No. MAB10971

Rat monoclonal antibody raised against Tek.

Specifications

Antigen Tek
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone TEK4
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Tek.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Gene Tek
Gene Alias AA517024/Cd202b/Hyk/Tie2/tie-2
Gene Symbols Tek
Host Species Rat
Immunogen Mouse Tek.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21687
Test Specificity The monoclonal antibody reacts with mouse Tek.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.