Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rat anti-TEL-AML1 fusion, Clone: 6F2,

Catalog No. MABE1007MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1007MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1007MI Supplier MilliporeSigma™ Supplier No. MABE1007
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects TEL-AML1 fusion clone: 6F2 in Human samples, and it is validated for Chromatin Immunoprecipitation, Flow Cytometry, Western Blotting

The chimeric transcription factor TEL-AML1 (ETV6-RUNX1) is the result of the reciprocal translocation t(12;21)(p13;q22), the most common structural genomic alteration found in childhood B-cell precursor acute lymphoblastic leukaemia (ALL). The runt-homology DNA-binding domain of AML1 retained in the TEL-AML1 mediates the DNA-binding activity of the fusion protein. The fusion protein not only exhibits altered transcription activity, it also affects normal transcription activity by directly competing against wild-type AML1 for promoter binding, sequestering transcriptional cofactors available for the wild-type protein, as well as dimerizing with wild-type protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEL-AML1 fusion
Applications ChIP Assay, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 6F2
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P41212; Q01196
Gene Symbols ETV6; TEL; TEL1; RUNX1; AML1; CBFA2
Host Species Rat
Immunogen KLH-conjugated linear peptide corresponding to human TEL-AML1 fusion.
Purification Method Protein G Purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001978; NP_001116079
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.