Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thy1, Rat, APC, Clone: G7, Abnova™

Catalog No. 89112581 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-581 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-581 Supplier Abnova Corporation Supplier No. MAB5956
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Thy1.

Specifications

Antigen Thy1
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone G7
Conjugate APC
Description Rat monoclonal antibody raised against Thy1.
Dilution Flow Cytometry (0.2 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Thy1
Gene Alias CD90/T25/Thy-1/Thy-1.2/Thy1.1/Thy1.2
Gene Symbols Thy1
Host Species Rat
Immunogen MST cell hybridoma C6/G8.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21838
Test Specificity Specificity Thy-1.1 and Thy-1.2 alloantigens (Thy-1 epitope region A)
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.