Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thy1, Rat, Clone: T-B8, Abnova™

Catalog No. 89110378 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-378 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-378 Supplier Abnova Corporation Supplier No. MAB1358
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native Thy1.

Specifications

Antigen Thy1
Applications Flow Cytometry
Classification Monoclonal
Clone T-B8
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native Thy1.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Thy1
Gene Alias CD90/T25/Thy-1/Thy-1.2/Thy1.1/Thy1.2
Gene Symbols Thy1
Host Species Rat
Immunogen B cell-depleted mouse LN cells., Native from Con A-activated
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21838
Test Specificity React with mouse Thy-1.
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.