Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat anti-TIM-1/KIM-1/HAVCR, Clone: RMT1-4, Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Specifications
Antigen | TIM-1/KIM-1/HAVCR |
---|---|
Clone | RMT1-4 |
Dilution | Flow Cytometry 1:10-1:1000 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14354105
![]() |
Novus Biologicals
NBP1435410.05MG |
0.05 mg | N/A | N/A | N/A | ||||
NBP1435415
![]() |
Novus Biologicals
NBP1435410.5MG |
0.5 mg | N/A | N/A | N/A | ||||
Description
TIM-1/KIM-1/HAVCR Monoclonal antibody specifically detects TIM-1/KIM-1/HAVCR in Mouse samples. It is validated for Flow Cytometry.Specifications
TIM-1/KIM-1/HAVCR | |
Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Rat | |
Mouse | |
HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
HAVCR1 | |
IgG2b κ | |
Affinity Purified |
RMT1-4 | |
Flow Cytometry | |
Biotin | |
RUO | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
26762 | |
The immunogen for this antibody was Purified TIM1. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title