Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tnfrsf11a, Rat, Clone: R12-31, Abnova™

Catalog No. 89113121 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-121 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-121 Supplier Abnova Corporation Supplier No. MAB7054

Rat monoclonal antibody raised against Tnfrsf11a.

Specifications

Antigen Tnfrsf11a
Applications Flow Cytometry
Classification Monoclonal
Clone R12-31
Conjugate Unconjugated
Description tumor necrosis factor receptor superfamily, member 11a
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Gene Tnfrsf11a
Gene Alias Ly109, ODFR, OFE, Rank, TRANCE-R, mRANK
Gene Symbols Tnfrsf11a
Host Species Rat
Immunogen Full length mouse Tnfrsf11a.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21934
Test Specificity RANK (R12-31).
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2a Kappa
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.