Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tnfrsf13b, Rat, Clone: 8F10, Abnova™

Catalog No. 89111569 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-569 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-569 Supplier Abnova Corporation Supplier No. MAB3730
Add to Cart
Add to Cart

Rat monoclonal antibody raised against partial recombinant Tnfrsf13b.

Specifications

Antigen Tnfrsf13b
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 8F10
Conjugate Unconjugated
Description Rat monoclonal antibody raised against partial recombinant Tnfrsf13b.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene Tnfrsf13b
Gene Alias 1200009E08Rik/Taci
Gene Symbols Tnfrsf13b
Host Species Rat
Immunogen Recombinant MBP fusion protein corresponding to extracellular region of mouse Tnfrsf13b.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57916
Test Specificity Recognizes mouse TACI.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.