Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tnfrsf17, Rat, Clone: Vicky-2, Abnova™

Catalog No. 89111381 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-381 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-381 Supplier Abnova Corporation Supplier No. MAB3374
Add to Cart
Add to Cart

Rat monoclonal antibody raised against partial recombinant Tnfrsf17.

Specifications

Antigen Tnfrsf17
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Vicky-2
Conjugate Unconjugated
Description Rat monoclonal antibody raised against partial recombinant Tnfrsf17.
Dilution Western Blots (1:1000) Immunoprecipitation (1:200) Flow Cytometry (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene Tnfrsf17
Gene Alias BCM/BCMA/Tnfrsf13/Tnfrsf13a
Gene Symbols Tnfrsf17
Host Species Rat
Immunogen Recombinant protein corresponding to chimeric protein with amino acids 1-46 of mouse Tnfrsf17 fused at the C-terminus to a 2 amino acid linker to the Fc portion of human IgG1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21935
Test Specificity Recognizes mouse BCMA. Does not cross-react with human BCMA.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.