Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tnfrsf18, Rat, Clone: DTA-1, Abnova™

Catalog No. 89112762 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-762 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-762 Supplier Abnova Corporation Supplier No. MAB6244
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native Tnfrsf18.

Specifications

Antigen Tnfrsf18
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone DTA-1
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native Tnfrsf18.
Dilution Flow Cytometry (0.1-0.5 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene Tnfrsf18
Gene Alias AITR/Gitr
Gene Symbols Tnfrsf18
Host Species Rat
Immunogen Native purified Tnfrsf18 from mouse CD25+ CD4+ T cell line.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21936
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.