Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFSF13, Rat, Clone: Sacha-1, Abnova™

Catalog No. 89111373 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-373 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-373 Supplier Abnova Corporation Supplier No. MAB3364
Add to Cart
Add to Cart

Rat monoclonal antibody raised against partial recombinant TNFSF13.

The protein encoded by this gene is a member of the tumor necrosis factor (TNF) ligand family. This protein is a ligand for TNFRSF17/BCMA, a member of the TNF receptor family. This protein and its receptor are both found to be important for B cell development. In vitro experiments suggested that this protein may be able to induce apoptosis through its interaction with other TNF receptor family proteins such as TNFRSF6/FAS and TNFRSF14/HVEM. Three alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. Other transcripts that skip the last exon of the upstream gene (TNFSF12) and continue with the second exon of this gene have been identified; such read-through transcripts are contained in GeneID 407977, TNFSF12-TNFSF13. [provided by RefSeq

Specifications

Antigen TNFSF13
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone Sacha-1
Conjugate Unconjugated
Description Rat monoclonal antibody raised against partial recombinant TNFSF13.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene TNFSF13
Gene Alias APRIL/CD256/TALL2/TRDL-1/UNQ383/PRO715/ligand
Gene Symbols TNFSF13
Host Species Rat
Immunogen Recombinant protein corresponding to chimeric protein with amino acids 105-250 of human TNFSF13l and amino acids 16-108 of human ACRP30 headless.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8741
Test Specificity Recognizes human and mouse APRIL.
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.