Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tnfsf4, Rat, Clone: MPG34, Abnova™

Catalog No. 89111564 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-564 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-564 Supplier Abnova Corporation Supplier No. MAB3725
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Tnfsf4.

Specifications

Antigen Tnfsf4
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MPG34
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Tnfsf4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene Tnfsf4
Gene Alias Ath-1/Ath1/CD134L/OX-40L/Ox40l/TXGP1/Txgp1l/gp34
Gene Symbols Tnfsf4
Host Species Rat
Immunogen Rat cells stably overexpressing mouse Tnfsf4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22164
Test Specificity Recognizes mouse CD134L. Does not cross-react with human CD134L.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.