Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tollip, Rat, Clone: Kimmy-1, Abnova™

Catalog No. 89111551 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-551 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-551 Supplier Abnova Corporation Supplier No. MAB3709
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant Tollip.

Specifications

Antigen Tollip
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Kimmy-1
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant Tollip.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene Tollip
Gene Alias 4930403G24Rik/4931428G15Rik
Gene Symbols Tollip
Host Species Rat
Immunogen Recombinant protein corresponding to mouse Tollip.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54473
Test Specificity Recognizes human and mouse Tollip.
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.