Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFB, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116724 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-724 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-724 Supplier Abnova Corporation Supplier No. PAB13528

Rat polyclonal antibody raised against recombinant Vegfb.

Specifications

Antigen VEGFB
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rat polyclonal antibody raised against recombinant Vegfb.
Dilution Western Blot (1-10 ug/mL) Immunohistochemistru (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from purified antibody without addition stabilizers.
Gene Vegfb
Gene Alias VEGF-B/Vrf
Gene Symbols Vegfb
Host Species Rat
Immunogen Recombinant protein corresponding to mouse Vegfb.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22340
Test Specificity This antibody reacts with murine VEGFB 186, no reactivity with VEGFB 167 in Western Blot.
Target Species Mouse
Content And Storage Store at -20°C on dry atmosphere.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.