Learn More
XRCC5, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of XRCC5.
Supplier: Abnova Corporation PAB24993
Description
The protein encoded by this gene is the 80-kilodalton subunit of the Ku heterodimer protein which is also known as ATP-dependant DNA helicase II or DNA repair protein XRCC5. Ku is the DNA-binding component of the DNA-dependent protein kinase, and it functions together with the DNA ligase IV-XRCC4 complex in the repair of DNA double-strand break by nonhomologous end joining and the completion of V(D)J recombination events. This gene functionally complements Chinese hamster xrs-6, a mutant defective in DNA double-strand break repair and in ability to undergo V(D)J recombination. A rare microsatellite polymorphism in this gene is associated with cancer in patients of varying radiosensitivity. [provided by RefSeq
Specifications
XRCC5 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of XRCC5. | |
In PBS, pH 7.2 (0.09% sodium azide) | |
FLJ39089/KARP-1/KARP1/KU80/KUB2/Ku86/NFIV | |
Rat | |
Antigen affinity purification | |
RUO | |
7520 | |
Human | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
XRCC5 | |
XRCC5 | |
A synthetic peptide corresponding to XRCC5. | |
100 μL | |
Primary | |
XRCC5 polyclonal antibody detects endogenous levels of XRCC5 protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only