Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat B7-1/CD80 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1214
This item is not returnable.
View return policy
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Rat samples. It is validated for Western Blot.Specifications
B7-1/CD80 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
AAB60503 | |
CD80 | |
Mouse myeloma cell line NS0-derived recombinant rat B7-1/CD80 Ile39-Gln248 Accession # AAB60503 | |
500 μg | |
Primary | |
Detects rat B7-1/CD80 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human B7-1, recombinant mouse (rm) B7-1, rmB7-2, recombinant rat B7-2, rmB7-H1, rmB7-H2, rmB7-H3, or rmPD-L2 is observed. | |
Rat | |
Purified |
Western Blot | |
210408 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Activation B7-1 antigen, B7, B71, BB1, CD28LG1, CD28LGB7-1 antigen), CD80, CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
941 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction