Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat B7-1/CD80 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1214SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1214SP 25 μg
AF1214 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1214SP Supplier R&D Systems Supplier No. AF1214SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

B7-1/CD80 Polyclonal specifically detects B7-1/CD80 in Rat samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen B7-1/CD80
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAB60503
Gene Alias Activation B7-1 antigen, B7, B71, BB1, CD28LG1, CD28LGB7-1 antigen), CD80, CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Gene Symbols CD80
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat B7-1/CD80 Ile39-Gln248 Accession # AAB60503
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 941
Test Specificity Detects rat B7-1/CD80 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse (rm) B7-1 is observed, 5% cross-reactivity with recombinant human B7-1 is observed, and less than 1% cross-reactivity with rmB7-H3, recombinant rat B7-2, rmB7-H1, and rmPD-L2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.