Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat B7-2/CD86 Antibody, R&D Systems™

Goat Polyclonal Antibody
$214.00 - $561.50
Specifications
Antigen | B7-2/CD86 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, ELISA |
Classification | Polyclonal |
Description
B7-2/CD86 Polyclonal specifically detects B7-2/CD86 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
B7-2/CD86 | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Polyclonal | |
Goat | |
Rat | |
Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86 | |
CD86 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, ELISA | |
Unconjugated | |
RUO | |
O35531 | |
942 | |
Mouse myeloma cell line NS0-derived recombinant rat B7-2/CD86 Val29-Ile250 Accession # O35531 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects rat B7-2/CD86 in ELISAs and Western blots. In sandwich immunoassays, less than 20% cross-reactivity with recombinant mouse (rm) B7-2 is observed and less than 0.2% cross-reactivity with recombinant rat B7-1, rmB7-1, and recombinant human B7-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title