Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat beta 2-Microglobulin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3864SP
This item is not returnable.
View return policy
Description
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Rat samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
beta 2-Microglobulin | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
P07151 | |
B2M | |
Mouse myeloma cell line NS0-derived recombinant rat beta 2-Microglobulin Ile21-Met119 Accession # P07151 | |
25 μg | |
Primary | |
Detects rat beta 2-Microglobulin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant rat alpha 2-Macroglobulin is observed. | |
Rat | |
Purified |
Flow Cytometry, CyTOF | |
747502 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
B2M, beta 2Microglobulin, beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
567 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction