Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat BOK Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF825
This item is not returnable.
View return policy
Description
BOK Polyclonal specifically detects BOK in Rat samples. It is validated for Western Blot.Specifications
BOK | |
Polyclonal | |
Unconjugated | |
Bcl2-L-9, BCL2L9bcl-2-related ovarian killer protein, Bcl-2-like protein 9, BCL2-related ovarian killer, BOKL, hBOK, MGC4631 | |
Rabbit | |
Affinity Purified | |
RUO | |
666 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.8 ug/mL | |
BOK | |
KLH-coupled rat BOK synthetic peptide ARLLRAGLSWSAPERASPAPGC | |
80 μg | |
Primary | |
Detects rat BOK in Western blots. | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction