Learn More
Specifications
Specifications
| Preparation Method | Ready-to-use lysates are especially prepared as positive controls for separation by SDS-PAGE and subsequent western blot analysis. Lysates are prepared in denaturing buffer WITHOUT dissociating agents (i.e. no 2-mercaptoethanol or dithiothreitol has been added). Heat lysate to 95 C for 5 minutes and rapidly cool. If dissociating conditions are desired, add reducing agent prior to heating. The recommended loading volume / lane is 10-20 μL depending on the size format of your gel. |
| Quantity | 500 μg |
| Purity | Multi-step |
| Host Species | Rat |
| Content And Storage | Store at -70°C. Avoid freeze/thaw cycles. |
| Product Line | Novus |
| Target | Brain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
