Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat C-Reactive Protein/CRP Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $667.00
Specifications
Antigen | C-Reactive Protein/CRP |
---|---|
Clone | 233539 |
Concentration | LYOPH |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB17441SP
![]() |
R&D Systems
MAB17441SP |
25 μg |
Each for $200.50
|
|
|||||
MAB17441
![]() |
R&D Systems
MAB17441 |
500 μg |
Each for $667.00
|
|
|||||
Description
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Rat samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
C-Reactive Protein/CRP | |
LYOPH | |
ELISA | |
Unconjugated | |
Mouse | |
Rat | |
P48199 | |
1401 | |
Mouse myeloma cell line NS0-derived recombinant rat C-Reactive Protein/CRP His20-Ser230 Accession # P48199 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects rat C-Reactive Protein/CRP in direct ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant mouse CRP, recombinant porcine CRP, recombinant human (rh) Pentraxin 2, or rhPentraxin 3 is observed. |
233539 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895 | |
CRP | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title