Learn More
Novus Biologicals™ Rat Carbonic Anhydrase IX/CA9 ELISA Kit (Chemiluminescence)
Shop All R&D Systems Products

Specifications
Specifications
| Assay Range | 12.5 to 800 pg/mL |
| Assay Sensitivity | 7.5 pg/mL |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Other Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Rat CA9 in samples. No s ignificant cross reactivity or interference between Rat CA9 and analogues was observed. |
| Target Species | Rat |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 768 |
| Gene Symbol | CA9 |
| Quantity | 1 Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.