Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat CCL20/MIP-3 alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB540SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB540SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB540SP Supplier R&D Systems Supplier No. MAB540SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCL20/MIP-3 alpha Monoclonal specifically detects CCL20/MIP-3 alpha in Rat samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CCL20/MIP-3 alpha
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 83225
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 20-40 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P97884.1
Gene Alias beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARC, LARCLiver and activation-regulated chemokine, MIP3 alpha, MIP-3 alpha, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38
Gene Symbols CCL20
Host Species Mouse
Immunogen E. coli-derived recombinant rat CCL20/MIP-3 alpha Ala26-Met96 Accession # P97884.1
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6364
Test Specificity Detects rat CCL20/MIP-3 alpha in ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.