Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat CCL26-like/Eotaxin-3-like Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3878
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3878 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3878 Supplier R&D Systems Supplier No. BAF3878
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CCL26-like/Eotaxin-3-like Polyclonal specifically detects CCL26-like/Eotaxin-3-like in Rat samples. It is validated for Western Blot.

Specifications

Antigen CCL26-like/Eotaxin-3-like
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. NP_001102958
Host Species Goat
Immunogen E. coli-derived recombinant rat CCL26-like/Eotaxin-3-like Thr24-Trp94 Accession # NP_001102958
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2911
Test Specificity Detects rat CCL26-like/Eotaxin-3-like in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant mouse CCL26-like is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.